N'-[1-Amino-2-(4-methoxyphenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058725-1G
058725-500MG
101915-578EA
376.92
USD
101915-578
101915-576
N'-[1-Amino-2-(4-methoxyphenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-(4-methoxyphenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester
Purity: 95+%
CAS: 1053655-77-8
MDL Number: MFCD10568183
Molecular Formula: C14H21N3O4
Molecular Weight: 295,34
Hazard: Irritant
Formula:
C₁₄H₂₁N₃O₄ MW: 295.34 g/mol |
MDL Number:
MFCD10568183 CAS Number: 1053655-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...