N'-[1-Amino-1-(3-fluoro-4-methylphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
Supplier: Matrix Scientific
058723-1G
058723-500MG
101915-568EA
426.15
USD
101915-568
101915-566
N'-[1-Amino-1-(3-fluoro-4-methylphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-1-(3-fluoro-4-methylphenyl)methylidene]hydrazinecarboxylic acid-tert-butyl ester
Purity: 95+%
CAS: 1053655-97-2
MDL Number: MFCD10568192
Molecular Formula: C13H18FN3O2
Molecular Weight: 267,31
Hazard: Irritant
Formula:
C₁₃H₁₈FN₃O₂ MW: 267.31 g/mol |
MDL Number:
MFCD10568192 CAS Number: 1053655-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...