2-Amino-5-chloropyridine-3-sulfonyl chloride ≥95%
Supplier: Matrix Scientific
058718-500MG
058718-1G
101915-550EA
587.69
USD
101915-550
101915-548
2-Amino-5-chloropyridine-3-sulfonyl chloride ≥95%
2-Amino-5-chloropyridine-3-sulfonyl chloride
Purity: 95+%
CAS: 163137-45-9
MDL Number: MFCD12402514
Molecular Formula: C5H4Cl2N2O2S
Molecular Weight: 227,07
Hazard: Corrosive
Formula:
C₅H₄Cl₂N₂O₂S MW: 227.07 g/mol Melting Pt: 117…119 °C |
MDL Number:
MFCD12402514 CAS Number: 163137-45-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...