N'-[1-Amino-2-(4-chlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%

Supplier: Matrix Scientific

058712-1G 058712-500MG
101915-524EA 284.62 USD
101915-524 101915-526
N'-[1-Amino-2-(4-chlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester ≥95%
N'-[1-Amino-2-(4-chlorophenoxy)ethylidene]hydrazinecarboxylic acid-tert-butyl ester

Purity: 95+%
CAS: 1053655-81-4
MDL Number: MFCD10568185
Molecular Formula: C13H18ClN3O3
Molecular Weight: 299,76
Hazard: Irritant

Formula: C₁₃H₁₈ClN₃O₃
MW: 299.76 g/mol
MDL Number: MFCD10568185
CAS Number: 1053655-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR