4-(Nitrooxy)butan-1-ol ≥95%

Supplier: Matrix Scientific

058688-500MG
101915-418EA 961.54 USD
101915-418
4-(Nitrooxy)butan-1-ol ≥95%
4-(Nitrooxy)butan-1-ol

Purity: 95%
CAS: 22911-39-3
MDL Number: MFCD18157664
Molecular Formula: C4H9NO4
Molecular Weight: 135,12
Hazard: Irritant

Formula: C₄H₉NO₄
MDL Number: MFCD18157664
CAS Number: 22911-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR