2,2-Dimethyl-4-oxo-7-[(trifluoromethane)sulfonylox y]-2,4-dihydro-1,3-benzodioxin-5-yltrifluoromethane ≥95%

Supplier: Matrix Scientific

058644-500MG 058644-1G
101915-216EA 804.62 USD
101915-216 101915-218
2,2-Dimethyl-4-oxo-7-[(trifluoromethane)sulfonylox y]-2,4-dihydro-1,3-benzodioxin-5-yltrifluoromethane ≥95%
2,2-Dimethyl-4-oxo-7-[(trifluoromethane)sulfonylox y]-2,4-dihydro-1,3-benzodioxin-5-yltrifluoromethane

Purity: 95%
CAS: 603044-07-1
MDL Number: MFCD18157650
Molecular Formula: C12H8F6O9S2
Molecular Weight: 474,31
Hazard: Irritant

Formula: C₁₂H₈F₆O₉S₂
MDL Number: MFCD18157650
CAS Number: 603044-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR