{3-[(Benzyloxy)carbonyl]-2,4-difluorophenyl}amine ≥95%
Supplier: Matrix Scientific
058620-500MG
058620-1G
101915-144EA
401.54
USD
101915-144
101915-146
{3-[(Benzyloxy)carbonyl]-2,4-difluorophenyl}amine ≥95%
{3-[(Benzyloxy)carbonyl]-2,4-difluorophenyl}amine
Purity:
CAS: 918523-44-1
MDL Number: MFCD18157654
Molecular Formula: C14H11F2NO2
Molecular Weight: 263,25
Hazard: Irritant
Formula:
C₁₄H₁₁F₂NO₂ |
MDL Number:
MFCD18157654 CAS Number: 918523-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...