1-(4-Aminophenyl)-5-methyl-1H-pyrazole-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
058612-500MG
058612-1G
101915-110EA
806.15
USD
101915-110
101915-112
1-(4-Aminophenyl)-5-methyl-1H-pyrazole-3-carboxylic acid ≥95%
1-(4-Aminophenyl)-5-methyl-1H-pyrazole-3-carboxylic acid
Purity:
CAS: 1261079-60-0
MDL Number: MFCD18157649
Molecular Formula: C11H11N3O2
Molecular Weight: 217,23
Hazard: Irritant
Formula:
C₁₁H₁₁N₃O₂ |
MDL Number:
MFCD18157649 CAS Number: 1261079-60-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...