4-(Pyrrolidin-3-yl)morpholine
Supplier: Matrix Scientific
Synonyms:
4-(3-Pyrrolidinyl)morpholine
058608-250MG
058608-1G
101915-098EA
261.13
USD
101915-098
101915-100
4-(Pyrrolidin-3-yl)morpholine
4-(Pyrrolidin-3-yl)morpholine
Hazard: Irritant
Formula:
C₈H₁₆N₂O Boiling Pt: 88…90 °C (0.6 mmHg) |
MDL Number:
MFCD06740352 CAS Number: 53617-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...