2-Chloro-3-fluoro-4-(difluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-(difluoromethyl)-3-fluoropyridine

058604-1G
101915-088EA 361.54 USD
101915-088
2-Chloro-3-fluoro-4-(difluoromethyl)pyridine ≥97%
2-Chloro-3-fluoro-4-(difluoromethyl)pyridine

Purity: 97%
CAS: 1262413-59-1
MDL Number: MFCD18384831
Molecular Formula: C6H3ClF3N
Molecular Weight: 181,55
Boiling Point: 70°C@ 20 mbar
Hazard: Irritant

Formula: C₆H₃ClF₃N
MW: 181.55 g/mol
Boiling Pt: 70 °C ( 20 mbar)
MDL Number: MFCD18384831
CAS Number: 1262413-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR