2-Chloro-3-fluoroisonicotinaldehyde ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3-fluoropyridine-4-carbaldehyde

058603-1G
101915-086EA 36.92 USD
101915-086
2-Chloro-3-fluoroisonicotinaldehyde ≥97%
2-Chloro-3-fluoroisonicotinaldehyde

Hazard: Irritant/Refrigerate

Formula: C₆H₃ClFNO
MW: 159.5455 g/mol
MDL Number: MFCD04972388
CAS Number: 329794-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR