5-(2-Bromoethyl)-2,2-dimethyl-1,3-dioxane ≥95%

Supplier: Matrix Scientific

058602-500MG
101915-084EA 400 USD
101915-084
5-(2-Bromoethyl)-2,2-dimethyl-1,3-dioxane ≥95%
5-(2-Bromoethyl)-2,2-dimethyl-1,3-dioxane

Purity: 95%
CAS: 97845-58-4
MDL Number: MFCD06411062
Molecular Formula: C8H15BrO2
Molecular Weight: 223,12
Hazard: Irritant-Harmful/Moisture Sensitive/Refrigerate

Formula: C₈H₁₅BrO₂
MDL Number: MFCD06411062
CAS Number: 97845-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR