Benzo[d]isoxazol-3-ol ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Hydroxy-1,2-benzisoxazole
058596-1G
058596-5G
101915-076EA
310.77
USD
101915-076
101915-074
Benzo[d]isoxazol-3-ol ≥98%
Benzo[d]isoxazol-3-ol
Purity: 98%
CAS: 21725-69-9
MDL Number: MFCD00125030
Molecular Formula: C7H5NO2
Molecular Weight: 135,12
Hazard: Irritant
Formula:
C₇H₅NO₂ MW: 135.12 g/mol |
MDL Number:
MFCD00125030 CAS Number: 21725-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...