2-Chloro-5-chlorosulfonyl-4-fluorobenzoic acid ≥98%
Supplier: Matrix Scientific
058559-5G
058559-25G
101915-054EA
300
USD
101915-054
101915-052
2-Chloro-5-chlorosulfonyl-4-fluorobenzoic acid ≥98%
2-Chloro-5-chlorosulfonyl-4-fluorobenzoic acid
Purity: 98%
CAS: 264927-50-6
MDL Number: MFCD01321365
Molecular Formula: C7H3Cl2FO4S
Molecular Weight: 273,07
Hazard: Irritant
Formula:
C₇H₃Cl₂FO₄S MW: 273.07 g/mol |
MDL Number:
MFCD01321365 CAS Number: 264927-50-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...