tert-Butyl-2-[3-(hydroxymethyl)-5-methyl-1H-pyrazol-1-yl]benzoate ≥95%
Supplier: Matrix Scientific
058512-1G
058512-5G
058512-500MG
101914-998EA
2670.77
USD
101914-998
101914-996
101914-994
tert-Butyl-2-[3-(hydroxymethyl)-5-methyl-1H-pyrazol-1-yl]benzoate ≥95%
tert-Butyl-2-[3-(hydroxymethyl)-5-methyl-1H-pyrazol-1-yl]benzoate
Purity:
CAS: 1251950-61-4
MDL Number: MFCD17392913
Molecular Formula: C16H20N2O3
Molecular Weight: 288,35
Hazard: Irritant
Formula:
C₁₆H₂₀N₂O₃ MW: 288.35 g/mol |
MDL Number:
MFCD17392913 CAS Number: 1251950-61-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...