Methyl-1-[2-(hydroxymethyl)phenyl]-5-methyl-1H-pyrazole-3-carboxylate ≥95%
Supplier: Matrix Scientific
058504-500MG
058504-5G
058504-1G
101914-964EA
2129.23
USD
101914-964
101914-962
101914-960
Methyl-1-[2-(hydroxymethyl)phenyl]-5-methyl-1H-pyrazole-3-carboxylate ≥95%
Methyl-1-[2-(hydroxymethyl)phenyl]-5-methyl-1H-pyrazole-3-carboxylate
Purity:
CAS: 1251950-60-3
MDL Number: MFCD17392911
Molecular Formula: C13H14N2O3
Molecular Weight: 246,27
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O₃ |
MDL Number:
MFCD17392911 CAS Number: 1251950-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...