Methyl-1-(2-aminophenyl)-5-methyl-1H-pyrazole-3-carboxylate ≥95%
Supplier: Matrix Scientific
058503-500MG
058503-5G
058503-1G
101914-958EA
2018.46
USD
101914-958
101914-956
101914-954
Methyl-1-(2-aminophenyl)-5-methyl-1H-pyrazole-3-carboxylate ≥95%
Methyl-1-(2-aminophenyl)-5-methyl-1H-pyrazole-3-carboxylate
Purity:
CAS: 1251950-62-5
MDL Number: MFCD17392912
Molecular Formula: C12H13N3O2
Molecular Weight: 231,26
Hazard: Irritant
Formula:
C₁₂H₁₃N₃O₂ |
MDL Number:
MFCD17392912 CAS Number: 1251950-62-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...