1,2,3,4-Naphthalenetetrone

Supplier: Matrix Scientific

058468-500MG
101914-868EA 306.15 USD
101914-868
1,2,3,4-Naphthalenetetrone
1,2,3,4-Naphthalenetetrone

CAS: 30266-58-1
MDL Number: MFCD00985549
Molecular Formula: C10H4O4
Molecular Weight: 188,14
Hazard: Irritant

Formula: C₁₀H₄O₄
MDL Number: MFCD00985549
CAS Number: 30266-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR