3-Hydroxyquinuclidine-3-carbonitrile

Supplier: Matrix Scientific

058462-500MG
101914-856EA 183.08 USD
101914-856
3-Hydroxyquinuclidine-3-carbonitrile
3-Hydroxyquinuclidine-3-carbonitrile

CAS: 6238-30-8
MDL Number: MFCD00673168
Molecular Formula: C8H12N2O
Molecular Weight: 152,2
Hazard: Irritant

Formula: C₈H₁₂N₂O
MDL Number: MFCD00673168
CAS Number: 6238-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR