4-Methoxy-7H-pyrimido[4,5-b][1,4]thiazin-6-amine

Supplier: Matrix Scientific

058458-500MG
101914-848EA 306.15 USD
101914-848
4-Methoxy-7H-pyrimido[4,5-b][1,4]thiazin-6-amine
4-Methoxy-7H-pyrimido[4,5-b][1,4]thiazin-6-amine

CAS: 37497-00-0
MDL Number: MFCD00807384
Molecular Formula: C7H8N4OS
Molecular Weight: 196,23
Hazard: Irritant

Formula: C₇H₈N₄OS
MDL Number: MFCD00807384
CAS Number: 37497-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR