8-Cyclohexyl-5,6-dihydro-4H-pyrazino[3,2,1-jk]carbazole
Supplier: Matrix Scientific
058449-500MG
101914-830EA
233.85
USD
101914-830
8-Cyclohexyl-5,6-dihydro-4H-pyrazino[3,2,1-jk]carbazole
8-Cyclohexyl-5,6-dihydro-4H-pyrazino[3,2,1-jk]carbazole
CAS: 157056-89-8
MDL Number: MFCD00409221
Molecular Formula: C20H22N2
Molecular Weight: 290,41
Hazard: Irritant
Formula:
C₂₀H₂₂N₂ MW: 290.41 g/mol |
MDL Number:
MFCD00409221 CAS Number: 157056-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...