N-(3-Methoxyphenyl)acetamide

Supplier: Matrix Scientific

058448-500MG
101914-828EA 306.15 USD
101914-828
N-(3-Methoxyphenyl)acetamide
N-(3-Methoxyphenyl)acetamide

CAS: 588-16-9
MDL Number: MFCD00123297
Molecular Formula: C9H11NO2
Molecular Weight: 165,19
Hazard: Irritant

Formula: C₉H₁₁NO₂
MDL Number: MFCD00123297
CAS Number: 588-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR