Methyl-4-methyl-3-[(N-propylalanyl)amino]-2-thiophenecarboxylate hydrochloride

Supplier: Matrix Scientific

058446-500MG
101914-824EA 306.15 USD
101914-824
Methyl-4-methyl-3-[(N-propylalanyl)amino]-2-thiophenecarboxylate hydrochloride
Methyl-4-methyl-3-[(N-propylalanyl)amino]-2-thiophenecarboxylate hydrochloride

CAS: 23964-57-0
MDL Number: MFCD00190157
Molecular Formula: C13H21ClN2O3S
Molecular Weight: 320,84
Hazard: Irritant

Formula: C₁₃H₂₁ClN₂O₃S
MDL Number: MFCD00190157
CAS Number: 23964-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR