Thiotepa
Supplier: Matrix Scientific
Synonyms:
Tri(aziridin-1-yl)phosphine sulfide
, Tri(1-aziridinyl)phosphine sulfide
, Tri(aziridin-1-yl)phosphine sulphide
, Tri(1-aziridinyl)phosphine sulphide
058443-500MG
101914-818EA
69.23
USD
101914-818
Thiotepa
Thiotepa
Hazard: Toxic
Formula:
C₆H₁₂N₃PS MW: 189.217 g/mol Melting Pt: 54…57 °C |
MDL Number:
MFCD00145452 CAS Number: 52-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...