Dicyclohexano-18-crown-6 (mixture of isomers)

Supplier: Matrix Scientific

Synonyms: Icosahydrodibenzo[b,k][1,4,7,10,13,16]-hexaoxacyclooctadecine

058442-500MG
101914-816EA 306.15 USD
101914-816
Dicyclohexano-18-crown-6 (mixture of isomers)
Dicyclohexano-18-crown-6 (mixture of isomers)

Hazard: Irritant

Formula: C₂₀H₃₆O₆
MDL Number: MFCD00005099
CAS Number: 16069-36-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR