Nifedipine
Supplier: Matrix Scientific
Synonyms:
1,4-Dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylic Acid Dimethyl Ester
, Dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
, Adalat
058437-500MG
101914-806EA
306.15
USD
101914-806
Nifedipine
Nifedipine
CAS: 21829-25-4
MDL Number: MFCD00057326
Molecular Formula: C17H18N2O6
Molecular Weight: 346,34
Hazard: Irritant
Formula:
C₁₇H₁₈N₂O₆ MW: 346.34 g/mol |
MDL Number:
MFCD00057326 CAS Number: 21829-25-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...