6,7-Dihydro-5H-pyrrolo[3,4-d]pyrimidine dihydrochloride
Supplier: Matrix Scientific
053868-500MG
053868-2.5G
101905-290EA
838.46
USD
101905-290
101905-288
6,7-Dihydro-5H-pyrrolo[3,4-d]pyrimidine dihydrochloride
6,7-Dihydro-5H-pyrrolo[3,4-d]pyrimidine dihydrochloride
Hazard: Irritant
Formula:
C₆H₉Cl₂N₃ |
MDL Number:
MFCD08457576 CAS Number: 157327-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...