3-Isopropylcyclohexanone

Supplier: Matrix Scientific

053817-2.5G 053817-500MG
101905-120EA 272.55 USD
101905-120 101905-122
3-Isopropylcyclohexanone
3-Isopropylcyclohexanone

Hazard: Irritant

Formula: C₉H₁₆O
CAS Number: 23396-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR