2,4-Diphenylpyrrolidine

053807-2.5G
101905-082EA 838.46 USD
101905-082
2,4-Diphenylpyrrolidine
2,4-Diphenylpyrrolidine

Hazard: Irritant

Formula: C₁₆H₁₇N
MDL Number: MFCD13562920
CAS Number: 39995-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR