2-Ethyl-1H-benzimidazole-6-carboxylic acid

Supplier: Matrix Scientific

053795-500MG 053795-2.5G
101905-036EA 364.62 USD
101905-036 101905-038
2-Ethyl-1H-benzimidazole-6-carboxylic acid
2-Ethyl-1H-benzimidazole-6-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD06799537
CAS Number: 891454-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR