2-(Methoxymethyl)benzoic acid

Supplier: Matrix Scientific

053785-2.5G 053785-500MG
101905-000EA 307.69 USD
101905-000 101905-002
2-(Methoxymethyl)benzoic acid
2-(Methoxymethyl)benzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₃
MDL Number: MFCD09863839
CAS Number: 88550-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR