4-(3-Pyridyl)benzaldehyde
Supplier: Matrix Scientific
Synonyms:
4-(Pyridin-3-yl)benzaldehyde
053770-500MG
053770-2.5G
101904-948EA
838.46
USD
101904-948
101904-946
4-(3-Pyridyl)benzaldehyde
4-(3-Pyridyl)benzaldehyde
Hazard: Irritant
Formula:
C₁₂H₉NO MW: 183.2060 g/mol |
MDL Number:
MFCD01631910 CAS Number: 127406-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...