Methyl-2-aminopyrimidine-5-carboxylate
Supplier: Matrix Scientific
Synonyms:
Methyl 2-aminopyrimidine-5-carboxylate
053744-500MG
053744-2.5G
101904-844EA
331.33
USD
101904-844
101904-842
Methyl-2-aminopyrimidine-5-carboxylate
Methyl-2-aminopyrimidine-5-carboxylate
Hazard: Irritant
Formula:
C₆H₇N₃O₂ MW: 153.1400 g/mol |
MDL Number:
MFCD08275693 CAS Number: 308348-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...