(Z)-3-(Dimethylamino)-2-formyl-2-propenenitrile

Supplier: Matrix Scientific

053732-500MG 053732-2.5G
101904-796EA 838.46 USD
101904-796 101904-794
(Z)-3-(Dimethylamino)-2-formyl-2-propenenitrile
(Z)-3-(Dimethylamino)-2-formyl-2-propenenitrile

Hazard: Irritant

Formula: C₆H₈N₂O
MDL Number: MFCD09054809
CAS Number: 13974-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR