3-Ethyl-2-(1H-pyrazol-1-yl)benzoic acid

Supplier: Matrix Scientific

053718-2.5G 053718-500MG
101904-742EA 364.62 USD
101904-742 101904-744
3-Ethyl-2-(1H-pyrazol-1-yl)benzoic acid
3-Ethyl-2-(1H-pyrazol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₂N₂O₂
MDL Number: MFCD09054736
CAS Number: 1214622-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR