3-(3-Methoxybenzyl)piperidine

Supplier: Matrix Scientific

053700-500MG 053700-2.5G
101904-672EA 838.46 USD
101904-672 101904-670
3-(3-Methoxybenzyl)piperidine
3-(3-Methoxybenzyl)piperidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD05863593
CAS Number: 766487-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR