3-(3-Phenylpropyl)piperidine

Supplier: Matrix Scientific

053692-2.5G 053692-500MG
101904-648EA 838.46 USD
101904-648 101904-646
3-(3-Phenylpropyl)piperidine
3-(3-Phenylpropyl)piperidine

Hazard: Irritant

Formula: C₁₄H₂₁N
MDL Number: MFCD13562906
CAS Number: 53295-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR