3-(3-Phenoxypropyl)piperidine

Supplier: Matrix Scientific

053691-500MG 053691-2.5G
101904-642EA 364.62 USD
101904-642 101904-644
3-(3-Phenoxypropyl)piperidine
3-(3-Phenoxypropyl)piperidine

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD13562905
CAS Number: 1219977-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR