3-Phenethylpiperidine

Supplier: Matrix Scientific

053688-2.5G 053688-500MG
101904-632EA 838.46 USD
101904-632 101904-630
3-Phenethylpiperidine
3-Phenethylpiperidine

Hazard: Irritant

Formula: C₁₃H₁₉N
MDL Number: MFCD06738886
CAS Number: 136423-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR