3-Ethylpiperidine hydrochloride
Supplier: Matrix Scientific
053683-500MG
053683-2.5G
101904-610EA
364.62
USD
101904-610
101904-612
3-Ethylpiperidine hydrochloride
3-Ethylpiperidine hydrochloride
Hazard: Irritant
Formula:
C₇H₁₆ClN MW: 149.67 g/mol Melting Pt: 142 °C |
MDL Number:
MFCD19442002 CAS Number: 13603-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...