3-(2-Methylbenzyl)piperidine
Supplier: Matrix Scientific
053679-2.5G
053679-500MG
101904-594EA
364.62
USD
101904-594
101904-596
3-(2-Methylbenzyl)piperidine
3-(2-Methylbenzyl)piperidine
Hazard: Irritant
Formula:
C₁₃H₁₉N MW: 189.3 g/mol |
MDL Number:
MFCD05863599 CAS Number: 955314-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...