3-Propylpyrrolidine hydrochloride

053657-2.5G
101904-516EA 1083.01 USD
101904-516
3-Propylpyrrolidine hydrochloride
3-Propylpyrrolidine hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClN
MDL Number: MFCD26936320
CAS Number: 116632-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR