3-[2-(2-Methoxyethoxy)ethyl]pyrrolidine
Supplier: Matrix Scientific
053654-500MG
053654-2.5G
101904-504EA
838.46
USD
101904-504
101904-502
3-[2-(2-Methoxyethoxy)ethyl]pyrrolidine
3-[2-(2-Methoxyethoxy)ethyl]pyrrolidine
Hazard: Irritant
Formula:
C₉H₁₉NO₂ |
MDL Number:
MFCD13562884 CAS Number: 1220030-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...