2-Ethoxy-5-propylbenzaldehyde

Supplier: Matrix Scientific

053641-2.5G 053641-500MG
101904-452EA 838.46 USD
101904-452 101904-450
2-Ethoxy-5-propylbenzaldehyde
2-Ethoxy-5-propylbenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD06247412
CAS Number: 883540-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR