5-(sec-Butyl)-2-ethoxybenzaldehyde

Supplier: Matrix Scientific

053622-500MG 053622-2.5G
101904-374EA 364.62 USD
101904-374 101904-376
5-(sec-Butyl)-2-ethoxybenzaldehyde
5-(sec-Butyl)-2-ethoxybenzaldehyde

Hazard: Irritant

Formula: C₁₃H₁₈O₂
MDL Number: MFCD06246079
CAS Number: 883514-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR