5-Isopropyl-2-propoxybenzaldehyde
Supplier: Matrix Scientific
053619-500MG
053619-2.5G
101904-364EA
838.46
USD
101904-364
101904-362
5-Isopropyl-2-propoxybenzaldehyde
5-Isopropyl-2-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₈O₂ |
MDL Number:
MFCD06247933 CAS Number: 883538-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...