4-(Cyclopropylmethoxy)[1,1'-biphenyl]-3-carbaldehyde
Supplier: Matrix Scientific
053614-2.5G
053614-500MG
101904-342EA
364.62
USD
101904-342
101904-344
4-(Cyclopropylmethoxy)[1,1'-biphenyl]-3-carbaldehyde
4-(Cyclopropylmethoxy)[1,1'-biphenyl]-3-carbaldehyde
Hazard: Irritant
Formula:
C₁₇H₁₆O₂ |
MDL Number:
MFCD06246722 CAS Number: 883523-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...