3-Ethyl-4-propoxybenzaldehyde

Supplier: Matrix Scientific

053600-500MG 053600-2.5G
101904-286EA 364.62 USD
101904-286 101904-288
3-Ethyl-4-propoxybenzaldehyde
3-Ethyl-4-propoxybenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD06247483
CAS Number: 883528-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR