4-Ethoxy-3-ethylbenzaldehyde
Supplier: Matrix Scientific
053599-2.5G
053599-500MG
101904-282EA
364.62
USD
101904-282
101904-284
4-Ethoxy-3-ethylbenzaldehyde
4-Ethoxy-3-ethylbenzaldehyde
Hazard: Irritant
Formula:
C₁₁H₁₄O₂ |
MDL Number:
MFCD06247340 CAS Number: 883536-96-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...