3-(sec-Butyl)-4-(cyclopropylmethoxy)benzaldehyde

Supplier: Matrix Scientific

053586-2.5G 053586-500MG
101904-232EA 1934.49 USD
101904-232 101904-230
3-(sec-Butyl)-4-(cyclopropylmethoxy)benzaldehyde
3-(sec-Butyl)-4-(cyclopropylmethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₅H₂₀O₂
MDL Number: MFCD06246070
CAS Number: 883514-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR